Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crypthophane E 95%, Thermo Scientific™

Catalog No. AC297100500
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC297100500 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC297100500 Supplier Thermo Scientific Chemicals Supplier No. 297100500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102683-14-7
Molecular Formula C57H60O12
Synonym Crypthophane collet

Specifications

Color White
Infrared Spectrum Authentic
Quantity 50 mg
Formula Weight 937.1
Percent Purity ≥94%
Physical Form Fine Crystalline Powder
Chemical Name or Material Crypthophane E, 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.