accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHNONHNSOONNCl

CAS RN 1027513-54-7

IUPAC Name: 8-[4-({4-[(4-chlorophenyl)methyl]piperazin-1-yl}sulfonyl)phenyl]-1-propyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 8-(4-{4-[(4-chlorophenyl)methyl]piperazin-1-ylsulfonyl}phenyl)-1-propyl-3,7-dihydropurine-2,6-dione
Molecular Weight (g/mol): 543.04
Molecular Formula: C25H27ClN6O4S
InChi Key: JQZJACVYMPKVDS-UHFFFAOYSA-N
SMILES: CCCN1C(=O)NC2=C(NC(=N2)C2=CC=C(C=C2)S(=O)(=O)N2CCN(CC3=CC=C(Cl)C=C3)CC2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)