accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)NONH2O

CAS RN 102767-28-2

IUPAC Name: (2R)-2-(2-oxopyrrolidin-1-yl)butanamide
Synonyms: levitiracetam levetiracetamum
Molecular Weight (g/mol): 170.21
Molecular Formula: C8H14N2O2
InChi Key: HPHUVLMMVZITSG-ZCFIWIBFSA-N
SMILES: CC[C@@H](N1CCCC1=O)C(N)=O
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)