CAS RN 10277-43-7
Lanthanum Nitrate TS, Ricca Chemical
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, 99.9% (REO)
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 IUPAC Name: lanthanum(3+);trinitrate;hexahydrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, REacton™, 99.999% (REO)
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, 98+%
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, REacton™, 99.99% (REO)
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis)
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Lanthanum Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Lanthanum (La) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Lanthanum Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Lanthanum Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Lanthanum(III) nitrate hexahydrate, Solstice
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 IUPAC Name: lanthanum(3+);trinitrate;hexahydrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Sigma Aldrich Lanthanum(III) nitrate hexahydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Lanthanum(III) nitrate hexahydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Lanthanum Nitrate (Crystalline/Laboratory), Fisher Chemical
CAS: 10277-43-7 Molecular Formula: H12LaN3O15 Molecular Weight (g/mol): 433.01 MDL Number: MFCD00149751 InChI Key: GJKFIJKSBFYMQK-UHFFFAOYSA-N Synonym: lanthanum iii nitrate hexahydrate,lanthanum nitrate hexahydrate,unii-kj5g6nyf2w,kj5g6nyf2w,lanthanum iii nitrate, hexahydrate 1:3:6,lanthanum nitrate, 6-hydrate,ksc909a4f,lanthanum nitrate hexahydrate,,lanthanum 3+ hexahydrate trinitronate PubChem CID: 16211478 IUPAC Name: lanthanum(3+) hexahydrate trinitrate SMILES: O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
- 1
- 2