Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) nitrate hexahydrate, Solstice

Catalog No. 6000885 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-008-85 500 g
60-008-84 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-008-85 Supplier Solstice Supplier No. 61520500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a.,≥99.0% (titration)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10277-43-7
Molecular Formula H12LaN3O15
Molecular Weight (g/mol) 433.01
MDL Number MFCD00149751
InChI Key GJKFIJKSBFYMQK-UHFFFAOYSA-N
Synonym lanthanum iii nitrate hexahydrate, lanthanum nitrate hexahydrate, unii-kj5g6nyf2w, kj5g6nyf2w, lanthanum iii nitrate, hexahydrate 1:3:6, lanthanum nitrate, 6-hydrate, ksc909a4f, lanthanum nitrate hexahydrate,, lanthanum 3+ hexahydrate trinitronate
PubChem CID 16211478
IUPAC Name lanthanum(3+);trinitrate;hexahydrate
SMILES O.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 126°C
Melting Point 40°C
Quantity 500 g
Assay Percent Range ≥99.0%
Linear Formula La(NO3)3 · 6H2O
UN Number UN1477
Solubility Information 1580 g/L in Water
Formula Weight 433.01g/mol
Percent Purity ≥99% (titration)
Grade ACS
Appearance Crystalline
Chemical Name or Material Lanthanum(III) nitrate hexahydrate
Show More Show Less
Hazard Statement P220-P261-P305 + P351 + P338
RTECSNumber OE5250000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.