CAS RN 1028141-88-9
IUPAC Name:
λ²-ruthenium(2+) bis(2,2'-bipyridine) hexafluoro-λ⁵-phosphanuide triphenylphosphane 4-aminobutanoate
Synonyms:
λ²-ruthenium(2+) bis(bipyridyl) triphenylphosphine 4-aminobutyrate hexafluorophosphate
Molecular Weight (g/mol):
922.81
Molecular Formula:
C42H39F6N5O2P2Ru
InChi Key:
SWFWLLYVSYURHZ-UHFFFAOYSA-M
SMILES:
[Ru++].NCCCC([O-])=O.F[P-](F)(F)(F)(F)F.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ RuBi-GABA
CAS: 1028141-88-9 Molecular Formula: C42H39F6N5O2P2Ru