accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3COHNHOONHHOOHNNHHNNH2ONHONH2NHONHHNNH2HNOHOHNOHONHOCH3H3CHNOHONHOCH3CH3HNONHOH2NNH2NHOHOHNOCH3H3CNHOHNONH2H2N

CAS RN 1030384-98-5

IUPAC Name: 2,6-diamino-N-[({1-[(1-{[1-({1-[(1-{[4-carbamimidamido-1-({5-[5-carbamimidamido-2-(2-{2-[2-(2-{2-[2-(2,6-diaminohexanamido)acetamido]-3-methylbutanamido}-3-hydroxypropanamido)-4-methylpentanamido]-3-hydroxypropanamido}-3-(4-hydroxyphenyl)propanamido)pentanamido]-1-carbamoylpentyl}carbamoyl)butyl]carbamoyl}-2-(4-hydroxyphenyl)ethyl)carbamoyl]-2-hydroxyethyl}carbamoyl)-3-methylbutyl]carbamoyl}-2-hydroxyethyl)carbamoyl]-2-methylpropyl}carbamoyl)methyl]hexanamide
Synonyms: 2,6-diamino-N-[({1-[(1-{[1-({1-[(1-{[4-carbamimidamido-1-({5-[5-carbamimidamido-2-(2-{2-[2-(2-{2-[2-(2,6-diaminohexanamido)acetamido]-3-methylbutanamido}-3-hydroxypropanamido)-4-methylpentanamido]-3-hydroxypropanamido}-3-(4-hydroxyphenyl)propanamido)pentanamido]-1-carbamoylpentyl}carbamoyl)butyl]carbamoyl}-2-(4-hydroxyphenyl)ethyl)carbamoyl]-2-hydroxyethyl}carbamoyl)-3-methylbutyl]carbamoyl}-2-hydroxyethyl)carbamoyl]-2-methylpropyl}carbamoyl)methyl]hexanamide
Molecular Weight (g/mol): 1927.29
Molecular Formula: C86H147N27O23
InChi Key: VUYRSKROGTWHDC-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)C(CO)NC(=O)C(NC(=O)CNC(=O)C(N)CCCCN)C(C)C)C(=O)NC(CO)C(=O)NC(CC1=CC=C(O)C=C1)C(=O)NC(CCCNC(N)=N)C(=O)NCCCCC(NC(=O)C(CCCNC(N)=N)NC(=O)C(CC1=CC=C(O)C=C1)NC(=O)C(CO)NC(=O)C(CC(C)C)NC(=O)C(CO)NC(=O)C(NC(=O)CNC(=O)C(N)CCCCN)C(C)C)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)