accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1031799-40-2

IUPAC Name: 2-amino-5-{[amino(hydrazinyl)methylidene]amino}pentanoic acid hydrochloride
Synonyms: 2-amino-5-{[amino(hydrazinyl)methylidene]amino}pentanoic acid hydrochloride
Molecular Weight (g/mol): 225.68
Molecular Formula: C6H16ClN5O2
InChi Key: YGVMJVSXFJRHGQ-UHFFFAOYNA-N
SMILES: Cl.NNC(N)=NCCCC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)