accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1034343-98-0

IUPAC Name: Graphene
Synonyms: methane collocarb carbene, 13c-labeled 13C-labeled carbene kosmos spheron magecol grosafe humenegro furnal
Molecular Weight (g/mol): —
Molecular Formula: Cx
InChi Key: —
SMILES: C
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)