Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Graphene Nanoplatelets 6-8nm(thick), 25micro m(wide), TCI America™
SDP

Catalog No. G04385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G04385G Supplier TCI America Supplier No. G04385G

Chemical Identifiers

CAS 1034343-98-0
Molecular Formula Cx
Molecular Weight (g/mol) 16.04
MDL Number MFCD00144065
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
Synonym methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
PubChem CID 297
ChEBI CHEBI:16183
IUPAC Name Graphene
SMILES C

Specifications

Color Black
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Graphene Nanoplatelets 6-8nm(thick), 25micro m(wide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.