CAS RN 103476-24-0
IUPAC Name:
hexapotassium 2,3,5-trihydroxy-4,6-bis(phosphonatooxy)cyclohexyl phosphate
Synonyms:
hexapotassium 2,3,5-trihydroxy-4,6-bis(phosphonatooxy)cyclohexyl phosphate
Molecular Weight (g/mol):
648.63
Molecular Formula:
C6H9K6O15P3
InChi Key:
DVOPWCOAGWTTEN-UHFFFAOYNA-H
SMILES:
[K+].[K+].[K+].[K+].[K+].[K+].OC1C(O)C(OP([O-])([O-])=O)C(OP([O-])([O-])=O)C(O)C1OP([O-])([O-])=O
1
–
1
of
1
results
1
R&D Systems™ D-myo-Inositol 1,4,5-trisphosphate, hexapotassium salt
CAS: 103476-24-0 Molecular Formula: C6H9K6O15P3