Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ D-myo-Inositol 1,4,5-trisphosphate, hexapotassium salt
GREENER_CHOICESDP

Catalog No. 14821R Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-821-R 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-821-R Supplier R&D Systems™ Supplier No. 1482/1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Ca2+ mobilizing second messenger

Important second messenger involved in Ca2+ mobilization from intracellular stores (EC50 = 0.1 μM). Formed from the enzymatic hydrolysis of phosphatidyl inositol-4,5-bisphosphate.

Chemical Identifiers

CAS 103476-24-0
Molecular Formula C6H9K6O15P3

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 648.64
Physical Form Solid
Molecular Weight (g/mol) 648.634
Chemical Name or Material D-myo-Inositol 1,4,5-trisphosphate, hexapotassium salt
Recommended Storage Store at 20°
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.