CAS RN 1037184-44-3
IUPAC Name:
4,7-dichloro-3-hydroxy-3-[2-(4-methoxyphenyl)-2-oxoethyl]-2,3-dihydro-1H-indol-2-one
Synonyms:
4,7-dichloro-3-hydroxy-3-[2-(4-methoxyphenyl)-2-oxoethyl]-1H-indol-2-one
Molecular Weight (g/mol):
366.19
Molecular Formula:
C17H13Cl2NO4
InChi Key:
HLXSCTYHLQHQDJ-UHFFFAOYNA-N
SMILES:
COC1=CC=C(C=C1)C(=O)CC1(O)C(=O)NC2=C(Cl)C=CC(Cl)=C12