accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 103745-07-9

IUPAC Name: 1-[(4-methylbenzenesulfonyl)oxy]butan-2-ol
Synonyms: 1-[(4-methylbenzenesulfonyl)oxy]butan-2-ol
Molecular Weight (g/mol): 244.31
Molecular Formula: C11H16O4S
InChi Key: UVSOURZMHXZMJW-UHFFFAOYNA-N
SMILES: CCC(O)COS(=O)(=O)C1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results