CAS RN 103878-83-7
IUPAC Name:
hydrogen N-(2-aminoethyl)-5-chloropyridine-2-carboxamide chloride
Synonyms:
hydrogen lazabemide chloride
Molecular Weight (g/mol):
236.1
Molecular Formula:
C8H11Cl2N3O
InChi Key:
JMFKTFLARGGXCC-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].NCCNC(=O)C1=CC=C(Cl)C=N1
1
–
1
of
1
results
1
R&D Systems™ Lazabemide hydrochloride
CAS: 103878-83-7 Molecular Formula: C8H11Cl2N3O