accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)OH(S)(S)O(S)H3C(S)H3C

CAS RN 1039-17-4

IUPAC Name: (1S,3aS,3bS,9aS,11aS)-1-hydroxy-1,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: (1S,3aS,3bS,9aS,11aS)-1-hydroxy-1,9a,11a-trimethyl-2H,3H,3aH,3bH,4H,5H,8H,9H,11H-cyclopenta[a]phenanthren-7-one
Molecular Weight (g/mol): 300.44
Molecular Formula: C20H28O2
InChi Key: BFMZZSBHQOCQFQ-HTDHLNIYSA-N
SMILES: C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)C3=CC[C@]12C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results