accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONH

CAS RN 104-63-2

IUPAC Name: 2-(benzylamino)ethan-1-ol
Synonyms: 2-(benzylamino)ethanol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: XNIOWJUQPMKCIJ-UHFFFAOYSA-N
SMILES: OCCNCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)