Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylethanolamine, 96%

Catalog No. AAA1686636
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1686636 500 g
AAA168660E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1686636 Supplier Thermo Scientific Chemicals Supplier No. A1686636
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 104-63-2 | C9H13NO | 151.21 g/mol

Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.

Solubility
Soluble in water 162 g/L (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents..
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-63-2
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002840
InChI Key XNIOWJUQPMKCIJ-UHFFFAOYSA-N
Synonym n-benzylethanolamine, 2-benzylamino ethanol, benzylaminoethanol, benzylethanolamine, ethanol, 2-phenylmethyl amino, 2-benzylamino ethan-1-ol, benzyl ethanolamine, n-benzyle ethanol amine, ethanol, 2-benzylamino, unii-48121ms9jm
PubChem CID 4348
IUPAC Name 2-(benzylamino)ethanol
SMILES OCCNCC1=CC=CC=C1

Specifications

Melting Point -30°C
Density 1.063
pH 11
Boiling Point 240°C
Flash Point 130°C (266°F)
Refractive Index 1.543
Quantity 500 g
Beilstein 775164
Solubility Information Soluble in water 162g/L (25°C).
Formula Weight 151.21
Percent Purity 96%
Chemical Name or Material N-Benzylethanolamine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-221-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.