accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10416-59-8

IUPAC Name: trimethylsilyl N-(trimethylsilyl)ethanimidate
Synonyms: bis(trimethylsilyl)acetamide
Molecular Weight (g/mol): 203.43
Molecular Formula: C8H21NOSi2
InChi Key: SIOVKLKJSOKLIF-UHFFFAOYSA-N
SMILES: CC(O[Si](C)(C)C)=N[Si](C)(C)C
Molecular Weight (g/mol) 
  • (11)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (5)
Physical Form 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (5)
  • (5)
  • (11)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)
  • (5)
  • (3)
  • (5)