Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,O-Bis(trimethylsilyl) acetamide, BSA, EMD-Millipore

Catalog No. M1096490010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1096490010 10 mL Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096490010 Supplier MilliporeSigma Supplier No. 1.09649.0010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10416-59-8
Molecular Formula C8H21NOSi2
Molecular Weight (g/mol) 203.43
MDL Number MFCD00008270
InChI Key SIOVKLKJSOKLIF-UHFFFAOYSA-N
Synonym BSA
PubChem CID 6913588
IUPAC Name trimethylsilyl N-(trimethylsilyl)ethanimidate
SMILES CC(O[Si](C)(C)C)=N[Si](C)(C)C

Specifications

Packaging Glass
Vapor Pressure 10 hPa (50°C)
Density Density: 0.83g/cm3 (20°C)
Linear Formula CH3C[NSi(CH3)3]OSi(CH3)3
Quantity 10 mL
Formula Weight 203.43
Chemical Name or Material N, O-Bis(trimethylsilyl) acetamide
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.