accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CH3CCH3H3CONNPOOI

CAS RN 1041634-69-8

IUPAC Name: (4-{4-[3-oxo-6'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-spiro[2-benzofuran-1,9'-xanthen]-3'-yl]piperazin-1-yl}butyl)triphenylphosphanium iodide
Synonyms: (4-{4-[3-oxo-6'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)spiro[2-benzofuran-1,9'-xanthen]-3'-yl]piperazin-1-yl}butyl)triphenylphosphanium iodide
Molecular Weight (g/mol): 954.69
Molecular Formula: C52H53BIN2O5P
InChi Key: JPCCULDYUUMFAD-UHFFFAOYNA-M
SMILES: [I-].CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1(OC(=O)C3=CC=CC=C13)C1=C(O2)C=C(C=C1)N1CCN(CCCC[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)