accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3CCH3SiCH3H3CCH3SiCH3H3CGe

CAS RN 104164-54-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 292.2
Molecular Formula: C9H27GeSi3
InChi Key: SYWSGAXJMRGDKW-UHFFFAOYSA-N
SMILES: [Ge].C[Si](C)C.C[Si](C)C.C[Si](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results