Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(trimethylsilyl)germanium Hydride 96%, Thermo Scientific™

Catalog No. AC338692500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC338692500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC338692500 Supplier Thermo Scientific Chemicals Supplier No. 338692500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104164-54-7
Molecular Formula C9H27GeSi3
Molecular Weight (g/mol) 292.2
InChI Key SYWSGAXJMRGDKW-UHFFFAOYSA-N
Synonym tris trimethylsilyl germane, me3si 3geh, germanium,trimethylsilicon, tris trimethylsilyl germanium hydride
PubChem CID 6364987
IUPAC Name germanium;trimethylsilicon
SMILES C[Si](C)C.C[Si](C)C.C[Si](C)C.[Ge]

Specifications

Density 0.93
Linear Formula [(CH3)3Si]3GeH
Quantity 250 mg
Formula Weight 293.19
Chemical Name or Material Tris(trimethylsilyl)germanium hydride, 96%
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.