accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHSOOONa

CAS RN 104484-71-1

IUPAC Name: sodium 3-[(naphthalen-1-yl)amino]propane-1-sulfonate
Synonyms: sodium 3-(naphthalen-1-ylamino)propane-1-sulfonate
Molecular Weight (g/mol): 287.31
Molecular Formula: C13H14NNaO3S
InChi Key: HSOPFPPLFWZKJN-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCCNC1=CC=CC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results