Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 3-(1-Naphthylamino)propanesulfonate 98.0+%, TCI America™
SDP

Catalog No. N04955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N04955G Supplier TCI America Supplier No. N04955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An amine-based building block used in chemical research.

Chemical Identifiers

CAS 104484-71-1
Molecular Formula C13H14NNaO3S
Molecular Weight (g/mol) 287.309
MDL Number MFCD00059520
InChI Key HSOPFPPLFWZKJN-UHFFFAOYSA-M
Synonym sodium 3-naphthalen-1-ylamino propane-1-sulfonate, sodium 3-1-naphthylamino propanesulfonate, n-1-naphthyl-3-aminopropanesulfonic acid sodium salt, sodium 3-1-naphthylamino-1-propanesulfonate, sodium 3-naphthalen-1-yl amino propane-1-sulfonate, 1-propanesulfonicacid,3-1-naphthalenylamino-,sodiumsalt 1:1, 1-propanesulfonic acid, 3-1-naphthalenylamino-, monosodium salt, ksc909a8j, sodium 3-1-naphthylamino propane-1-sulfonate, n-1-naphthyl-3-aminopropanesulfonic acidsodiumsalt
PubChem CID 23672139
IUPAC Name sodium;3-(naphthalen-1-ylamino)propane-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2NCCCS(=O)(=O)[O-].[Na+]

Specifications

Quantity 5 g
Formula Weight 287.31
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-(1-Naphthylamino)propanesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.