accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104561-41-3

IUPAC Name: 4-[2-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethynyl]benzoic acid
Synonyms: 4-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethynyl]benzoic acid
Molecular Weight (g/mol): 332.44
Molecular Formula: C23H24O2
InChi Key: OQVLOWLEEHYBJH-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=C1C=CC(=C2)C#CC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results