accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10476-81-0

BrBrSr

CAS RN 10476-81-0

IUPAC Name: strontium(2+) dibromide
Synonyms: strontium bromide
Molecular Weight (g/mol): 247.43
Molecular Formula: Br2Sr
InChi Key: YJPVTCSBVRMESK-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Sr++]
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)