accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH2H2N

CAS RN 104821-25-2

IUPAC Name: 5-ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine
Synonyms: 5-ethyl-6-phenyl-6H-phenanthridine-3,8-diamine
Molecular Weight (g/mol): 315.42
Molecular Formula: C21H21N3
InChi Key: XYJODUBPWNZLML-UHFFFAOYNA-N
SMILES: CCN1C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results