accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONCl(E)

CAS RN 104830-08-2

IUPAC Name: ethyl (2E)-3-(2-chloropyridin-3-yl)prop-2-enoate
Synonyms: ethyl (2E)-3-(2-chloropyridin-3-yl)prop-2-enoate
Molecular Weight (g/mol): 211.65
Molecular Formula: C10H10ClNO2
InChi Key: AXJQCHYUAHBXAK-AATRIKPKSA-N
SMILES: CCOC(=O)\C=C\C1=CC=CN=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results