CAS RN 10486-08-5
CAS RN 10486-08-5
IUPAC Name:
sodium (4-methylphenyl)sulfanide
Synonyms:
sodium (4-methylphenyl)sulfanide
Molecular Weight (g/mol):
146.18
Molecular Formula:
C7H7NaS
InChi Key:
PRORQUQEDGGDQO-UHFFFAOYSA-M
SMILES:
[Na+].CC1=CC=C([S-])C=C1
1
–
2
of
2
results
1
Promotions Available
Sigma Aldrich 4-Methylbenzenethiol sodium salt
Sigma Aldrich 4-Methylbenzenethiol sodium salt
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.