CAS RN 1049731-36-3
IUPAC Name:
N-[3-({4-[(3-aminopropyl)amino]butyl}amino)propyl]-2-(naphthalen-1-yl)acetamide trihydrochloride
Synonyms:
N-[3-({4-[(3-aminopropyl)amino]butyl}amino)propyl]-2-(naphthalen-1-yl)acetamide trihydrochloride
Molecular Weight (g/mol):
479.92
Molecular Formula:
C22H37Cl3N4O
InChi Key:
JNEOJAUJWOPWJS-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.NCCCNCCCCNCCCNC(=O)CC1=CC=CC2=CC=CC=C12
1
–
1
of
1
results
1
R&D Systems™ Naspm trihydrochloride
CAS: 1049731-36-3 Molecular Formula: C22H37Cl3N4O