accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 105-99-7

IUPAC Name: 1,6-dibutyl hexanedioate
Synonyms: dibutyl adipate 3PS polycizer W 260 experimental tick repellent 3 experimental tick repellent 3PS dibutylester kyseliny adipove unitolate B cetiol B
Molecular Weight (g/mol): 258.36
Molecular Formula: C14H26O4
InChi Key: XTJFFFGAUHQWII-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CCCCC(=O)OCCCC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)