Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutyl Adipate, MP Biomedicals

Catalog No. ICN212990 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN212990 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN212990 Supplier MP Biomedicals, Inc Supplier No. 0521299005

Chemical Identifiers

CAS 105-99-7
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.36
MDL Number MFCD00009451
InChI Key XTJFFFGAUHQWII-UHFFFAOYSA-N
Synonym dibutyl adipate, butyl adipate, di-n-butyl adipate, hexanedioic acid, dibutyl ester, cetiol b, polycizer w 260, adipic acid, dibutyl ester, unitolate b, experimental tick repellent 3, di-butyl adipate
PubChem CID 7784
ChEBI CHEBI:34686
IUPAC Name 1,6-dibutyl hexanedioate
SMILES CCCCOC(=O)CCCCC(=O)OCCCC

Specifications

Quantity 100 g
Formula Weight 258.36
Chemical Name or Material Dibutyl Adipate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.