accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 10519-17-2

IUPAC Name: ethyl undec-2-ynoate
Synonyms: ethyl undec-2-ynoate
Molecular Weight (g/mol): 210.32
Molecular Formula: C13H22O2
InChi Key: WVNHQHYIPXYFQX-UHFFFAOYSA-N
SMILES: CCCCCCCCC#CC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results