accessibility menu, dialog, popup

UserName

Viewing profile as user:
KCH3SiCH3CH3HO

CAS RN 10519-96-7

IUPAC Name: trimethylsilanol potassium
Synonyms: trimethylsilanol potassium
Molecular Weight (g/mol): 129.3
Molecular Formula: C3H10KOSi
InChi Key: COTHYYYVPUZALV-UHFFFAOYSA-N
SMILES: [K].C[Si](C)(C)O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)