Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium trimethylsilanolate, 2M solution in THF, AcroSeal™

Catalog No. AC448468000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC448468000 800 mL AcroSeal™ Glass bottle
AC448461000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC448468000 Supplier Thermo Scientific Chemicals Supplier No. 448468000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 10519-96-7 | C3H10KOSi | 129.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10519-96-7 , 109-99-9
Molecular Formula C3H10KOSi
Molecular Weight (g/mol) 129.30
MDL Number MFCD00013421
InChI Key COTHYYYVPUZALV-UHFFFAOYSA-N
Synonym potassium trimethylsilanolate, trimethylsiloxypotassium, silanol, trimethyl-, potassium salt, trimethylsilanol potassium salt, potassium trimethyl silanolate, silanol, 1,1,1-trimethyl-, potassium salt 1:1, kosime3, kotms, tmsok, c3h9osi.k
PubChem CID 23668039
SMILES [K].C[Si](C)(C)O

Specifications

Quantity 800 mL
Linear Formula (CH3)3SiOK
Flash Point −17°C
IUPAC Name trimethylsilanol potassium
Formula Weight 128.29
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.91
Density 0.9100g/mL
Chemical Name or Material Potassium trimethylsilanolate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May form explosive peroxides.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 234-062-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.