CAS RN 1052087-90-7
IUPAC Name:
1-amino-9,10-dioxo-4-{[4-(phenylamino)-3-sulfophenyl]amino}-9,10-dihydroanthracene-2-sulfonic acid disodium
Synonyms:
1-amino-9,10-dioxo-4-{[4-(phenylamino)-3-sulfophenyl]amino}anthracene-2-sulfonic acid disodium
Molecular Weight (g/mol):
611.55
Molecular Formula:
C26H19N3Na2O8S2
InChi Key:
CVHGFNQCVCSUDU-UHFFFAOYSA-N
SMILES:
[Na].[Na].NC1=C(C=C(NC2=CC(=C(NC3=CC=CC=C3)C=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O