accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOON

CAS RN 10531-13-2

IUPAC Name: 2-[(benzenesulfonyl)oxy]acetonitrile
Synonyms: 2-[(benzenesulfonyl)oxy]acetonitrile
Molecular Weight (g/mol): 197.21
Molecular Formula: C8H7NO3S
InChi Key: SSOVHFPWMPUQOS-UHFFFAOYSA-N
SMILES: O=S(=O)(OCC#N)C1=CC=CC=C1
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results