Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyanomethyl Benzenesulfonate (Tech.), 80%, Thermo Scientific™

Catalog No. AC163060250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC163060250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC163060250 Supplier Thermo Scientific Chemicals Supplier No. 163060250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10531-13-2
Molecular Formula C8H7NO3S
Molecular Weight (g/mol) 197.208
InChI Key SSOVHFPWMPUQOS-UHFFFAOYSA-N
Synonym cyanomethylbenzenesulfonate, acetonitrile, phenylsulfonyl oxy, 2-benzenesulfonyl oxy acetonitrile, cyanomethyl benzenesulphonate, acmc-20aldf, phenylsulfonyloxyacetonitrile, benzenesulfonyloxyacetonitrile, phenylsulfonyl oxy acetonitrile, acetonitrile,2-phenylsulfonyl oxy
PubChem CID 82705
IUPAC Name cyanomethyl benzenesulfonate
SMILES C1=CC=C(C=C1)S(=O)(=O)OCC#N

Specifications

Linear Formula C6H5SO3CH2CN
Quantity 25 g
Formula Weight 197.21
Grade Technical
Chemical Name or Material Cyanomethyl benzenesulfonate, tech., 80%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.