accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105379-24-6

IUPAC Name: 1-[(1H-1,2,3-benzotriazol-1-yloxy)(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-[(1,2,3-benzotriazol-1-yloxy)(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium hexafluorophosphate
Molecular Weight (g/mol): 431.32
Molecular Formula: C15H20F6N5OP
InChi Key: XKTRAGMCMJYRRN-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.C1CCN(C1)C(ON1N=NC2=CC=CC=C12)=[N+]1CCCC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)