accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10553-31-8

IUPAC Name: barium(2+) dibromide
Synonyms: barium bromide
Molecular Weight (g/mol): 297.14
Molecular Formula: BaBr2
InChi Key: NKQIMNKPSDEDMO-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Ba++]
Percent Purity 
  • (4)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)