Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium bromide, 99%, pure, anhydrous

Catalog No. AC315380500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC315380500 50 g
AC315382500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC315380500 Supplier Thermo Scientific Chemicals Supplier No. 315380500
Add to Cart

missing translation for 'validMainframeMsg'

For ultra trace analysis | CAS: 10553-31-8 | BaBr2 | 297.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10553-31-8
Molecular Formula BaBr2
Molecular Weight (g/mol) 297.16

Specifications

Melting Point 850.0°C
Quantity 50 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula BaBr2
Merck Index 15,964
Solubility Information Solubility in water: 104.1g/100mL (20°C). Other solubilities: soluble in methanol, almost insoluble in ethanol, ethyl acetate,, acetone, dioxane
Formula Weight 297.16
Percent Purity 99%
Grade Pure
Chemical Name or Material Barium bromide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
EINECSNumber 234-140-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.