Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium bromide, ultra dry, 99.998% (metals basis)

Catalog No. AA3567803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3567803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3567803 Supplier Thermo Scientific Chemicals Supplier No. 03567803
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10553-31-8

Barium bromide is a precursor to chemicals used in photography and to other bromides. Barium bromide was used to purify radium in a process of fractional crystallization devised by Marie Curie. Since radium precipitates preferentially in a solution of barium bromide, the ratio of radium to barium in the precipitate would be higher than the ratio in the solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10553-31-8
MDL Number MFCD00003444

Specifications

Melting Point 850°C
Color White
Physical Form Beads
Quantity 1 g
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.781 g/mL
UN Number UN1564
Merck Index 14,968
Solubility Information It is soluble in water.
Formula Weight 297.14
Percent Purity 99.998%
Assay (metals basis)
Chemical Name or Material Barium bromide, Ultra dry
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
EINECSNumber 234-140-0
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.