accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOCH3NH2H2NHClHCl

CAS RN 1055301-09-1

IUPAC Name: methyl 4-{1,2-diamino-2-[4-(methoxycarbonyl)phenyl]ethyl}benzoate dihydrochloride
Synonyms: methyl 4-{1,2-diamino-2-[4-(methoxycarbonyl)phenyl]ethyl}benzoate dihydrochloride
Molecular Weight (g/mol): 401.28
Molecular Formula: C18H22Cl2N2O4
InChi Key: PABGVTNAPKOUBZ-UHFFFAOYNA-N
SMILES: Cl.Cl.COC(=O)C1=CC=C(C=C1)C(N)C(N)C1=CC=C(C=C1)C(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results