Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-1,2-Bis(4-methoxycarbonylphenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™

Catalog No. AAH25968MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25968MD 250 mg
AAH2596803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25968MD Supplier Thermo Scientific Chemicals Supplier No. H25968MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1055301-09-1
Molecular Formula C18H22Cl2N2O4
Molecular Weight (g/mol) 401.284
MDL Number MFCD09265307
InChI Key PABGVTNAPKOUBZ-UWGSCQAASA-N
Synonym r,r-1,2-bis 4-methoxycarbonylphenyl-1,2-ethanediamine dihydrochloride, ee, methyl 4-1r,2r-1,2-diamino-2-4-methoxycarbonyl phenyl ethyl benzoate dihydrochloride
PubChem CID 121233654
IUPAC Name methyl 4-[(1R,2R)-1,2-diamino-2-(4-methoxycarbonylphenyl)ethyl]benzoate;dihydrochloride
SMILES COC(=O)C1=CC=C(C=C1)C(C(C2=CC=C(C=C2)C(=O)OC)N)N.Cl.Cl

Specifications

Melting Point >200°C
Quantity 250 mg
Optical Rotation 47° (c=1 Methanol)
Formula Weight 401.3
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (R,R)-1,2-Bis(4-methoxycarbonylphenyl)-1,2-ethanediamine dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.