CAS RN 1055301-15-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R,R)-1,2-Bis(4-dimethylaminophenyl)-1,2-ethanediamine tetrahydrochloride, 95%, ee 99%, Thermo Scientific™
CAS: 1055301-15-9 Molecular Formula: C18H26N4·4HCl MDL Number: MFCD09265297