accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105816-04-4

IUPAC Name: (2R)-3-phenyl-2-{[(1S,3S)-3-(propan-2-yl)cyclohexyl]formamido}propanoic acid
Synonyms: nateglinide
Molecular Weight (g/mol): 317.43
Molecular Formula: C19H27NO3
InChi Key: CZCVBLROPSHDRZ-YESZJQIVSA-N
SMILES: CC(C)[C@H]1CCC[C@@H](C1)C(=O)N[C@H](CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)