accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-00-3

IUPAC Name: 1,6-bis(2-methoxyethyl) hexanedioate
Synonyms: 1,6-bis(2-methoxyethyl) hexanedioate
Molecular Weight (g/mol): 262.3
Molecular Formula: C12H22O6
InChi Key: GVRNUDCCYWKHMV-UHFFFAOYSA-N
SMILES: COCCOC(=O)CCCCC(=O)OCCOC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)