Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-methoxyethyl) adipate, 98%, Thermo Scientific™

Catalog No. AAA1049714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1049714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1049714 Supplier Thermo Scientific Chemicals Supplier No. A1049714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-00-3
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.302
MDL Number MFCD00041905
InChI Key GVRNUDCCYWKHMV-UHFFFAOYSA-N
Synonym bis 2-methoxyethyl adipate, hexanedioic acid, bis 2-methoxyethyl ester, bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate, adipic acid, bis 2-methoxyethyl ester, adipic acid bis 2-methoxyethyl ester, hexanedioic acid, 1,6-bis 2-methoxyethyl ester, 1,6-bis 2-methoxyethyl hexanedioate
PubChem CID 66046
IUPAC Name bis(2-methoxyethyl) hexanedioate
SMILES COCCOC(=O)CCCCC(=O)OCCOC

Specifications

Boiling Point 190°C (11 mmHg)
Refractive Index 1.4425
Linear Formula (CH2)4(CO2CH2CH2OCH3)2
Quantity 25 g
Formula Weight 262.3
Percent Purity 98%
Chemical Name or Material Bis (2-methoxyethyl) adipate
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 203-352-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.