CAS RN 106-96-7
Propargyl Bromide (stabilized with MgO) 97.0+%, TCI America™
CAS: 106-96-7 Molecular Formula: C3H3Br Molecular Weight (g/mol): 118.961 MDL Number: MFCD00000241 InChI Key: YORCIIVHUBAYBQ-UHFFFAOYSA-N Synonym: propargyl bromide,3-bromopropyne,3-bromo-1-propyne,1-propyne, 3-bromo,2-propynyl bromide,propynyl bromide,1-bromo-2-propyne,propyne, 3-bromo,gamma-bromoallylene,1-brom-2-propin PubChem CID: 7842 IUPAC Name: 3-bromoprop-1-yne SMILES: C#CCBr
Propargyl bromide, 80 wt.% solution in toluene, stabilized
CAS: 106-96-7 | C3H3Br | 118.961 g/mol
Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™
CAS: 106-96-7 | C3H3Br | 118.96 g/mol
Propargyl bromide, 97%, 80% w/w in toluene, stab. with magnesium oxide
CAS: 106-96-7 Molecular Formula: C3H3Br Molecular Weight (g/mol): 118.961 MDL Number: MFCD00000241 InChI Key: YORCIIVHUBAYBQ-UHFFFAOYSA-N Synonym: propargyl bromide,3-bromopropyne,3-bromo-1-propyne,1-propyne, 3-bromo,2-propynyl bromide,propynyl bromide,1-bromo-2-propyne,propyne, 3-bromo,gamma-bromoallylene,1-brom-2-propin PubChem CID: 7842 IUPAC Name: 3-bromoprop-1-yne SMILES: C#CCBr
Propargyl bromide, 80% in toluene, stab. with MgO
CAS: 106-96-7 Molecular Formula: C3H3Br Molecular Weight (g/mol): 118.961 MDL Number: MFCD00000241 InChI Key: YORCIIVHUBAYBQ-UHFFFAOYSA-N Synonym: propargyl bromide,3-bromopropyne,3-bromo-1-propyne,1-propyne, 3-bromo,2-propynyl bromide,propynyl bromide,1-bromo-2-propyne,propyne, 3-bromo,gamma-bromoallylene,1-brom-2-propin PubChem CID: 7842 IUPAC Name: 3-bromoprop-1-yne SMILES: C#CCBr
Propargyl Bromide (80% in Toluene, ca. 9.2mol/L) (stabilized with MgO), TCI America™
CAS: 106-96-7 Molecular Formula: C3H3Br Molecular Weight (g/mol): 118.961 MDL Number: MFCD00000241 InChI Key: YORCIIVHUBAYBQ-UHFFFAOYSA-N Synonym: propargyl bromide,3-bromopropyne,3-bromo-1-propyne,1-propyne, 3-bromo,2-propynyl bromide,propynyl bromide,1-bromo-2-propyne,propyne, 3-bromo,gamma-bromoallylene,1-brom-2-propin PubChem CID: 7842 IUPAC Name: 3-bromoprop-1-yne SMILES: C#CCBr
Sigma Aldrich Propargyl bromide solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Propargyl bromide solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.